Orders LinkContactUsLinkComponent
Intervoid® tamper evident mailing bags
Intervoid® tamper evident mailing bags
Intervoid®, tamper evident mailing bag with detachable receipt, 310×460 mm
Katalog # 129-1030
Leverandør:   COVERIS
Intervoid® tamper evident mailing bags
Katalog # 129-1030
Leverandør:  COVERIS
Intervoid® tamper evident mailing bags
Intervoid® tamper evident mailing bags
Intervoid®, tamper evident mailing bag with detachable receipt, 310×460 mm
Price
4.890,00 kr
Log ind for tilgængelighed
1 * 500 Items
Product Details & Documents
Intervoid® tamper evident mailing bags are designed to ensure the secure dispatch of samples or confidential data by mail courier. These respond to the critical needs of having a real guaranty of traceability, confidentiality and security when transporting samples.
  • Opaque polyethylene film protects the shipment from light and provides confidentiality
  • Traceability: one barcode with sequential numbering and detachable receipt
  • High-security: closure, unique marking system (pat. Pend.), micro printing
Specifikationer
  • Beskrivelse:
    Intervoid®, tamper evident mailing bag with detachable receipt
  • L×B:
    310×460 mm
  • Seq:
    03