Disinfectants for use in Ultrasonic baths, STAMMOPUR DR 8
Katalog # BAND972
Leverandør: BANDELIN-ELECTRONIC
Specifikationer
- Beskrivelse:STAMMOPUR DR 8, disinfectant and cleaning concentrate
- Pakn:2 L
- Til:SUPER, DIGITEC, DIGITAL, LONGLIFE
Specifikationer
Om denne vare
STAMMOPUR DR 8 concentrate is designed for cleaning of highly sensitive instruments using short sonication times in ultrasonic baths.
- Rapid disinfection and efficient cleaning in ultrasonic baths
- Formulation is free of aldehudes, phenols and chlorine
- Broad range spectrum of activity- bactericidal, tuberculocidal, yeasticidal
- Virucidal against BVDV, Vaccinia, Papova, HBV, HCV, HIV, A-H5N1
- High material compatibility: Metal, glass, porcelain, plastics and rubber