Orders LinkContactUsLinkComponent
Swing over bench
Swing over bench
Swing over bench, 850×350×450 mm
Katalog # 134-0501
Leverandør:   Palbam
Swing over bench
Katalog # 134-0501
Leverandør:  Palbam
Swing over bench
Swing over bench
Swing over bench, 850×350×450 mm
Price
10.850,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This bench is specifically designed to assist in putting on over shoes - by supporting the foot with a small raised platform. Two or more benches can be connected together (either seat-to-seat or foot support-to-foot support) to create the ideal step over bench.
  • The bench can also be used as a separation barrier between pre-gowning and gowning areas
  • Optional connection pieces allow benches to be joined together (see accessories)
Stainless steel 304 with electropolished finish.
Leveringsinformation: Typically available from stock for direct shipment from the supplier.
Specifikationer
  • Beskrivelse:
    Swing over bench
  • B×D×H:
    850×350×450 mm
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Bench corner connector 90˚
    Bench corner connector 90˚
    134-0481
    Log ind for tilgængelighed
    1 * 1 Items - kr1,380.00
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Bench corner connector 45˚
    Bench corner connector 45˚
    134-0480
    Log ind for tilgængelighed
    1 * 1 Items - kr1,730.00
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Bench corner connector 180˚
    Bench corner connector 180˚
    134-0482
    Log ind for tilgængelighed
    1 * 1 Items - kr1,110.00