Safety spectacles, Virtua™ AP
Leverandør: 3M
Gå til:
Om denne vare
Lightweight, stylish designed safety glasses Virtua™ AP protective eyewear features a streamline wrap-around design for excellent coverage and good field of vision.
- Integrated brow bar for added protection
- Lightweight for maximum comfort
- Strong polycarbonate lenses featuring anti-scratch coating for improved vision and extended durability
Weight: 26 g
Certifications: EN 166:2001
2 tilgængelige muligheder nedenfor
Product Details & Documents
Lightweight, stylish designed safety glasses Virtua™ AP protective eyewear features a streamline wrap-around design for excellent coverage and good field of vision.
- Integrated brow bar for added protection
- Lightweight for maximum comfort
- Strong polycarbonate lenses featuring anti-scratch coating for improved vision and extended durability
Weight: 26 g
Certifications: EN 166:2001