LC columns, MAbPac™ HIC-10
Om denne vare
The MAbPac™ HIC-10 column is based on high-purity, spherical, wide-pore silica particles functionalised with proprietary alkyl amide groups. Its advanced surface bonding technology leads to excellent chemical stability, high efficiency and unique selectivity optimised for MAbs and different from other HIC phases.
- Excellent biocompatibility and extremely low carryover
- High resolution and high efficiency
- Compatible with both organic solvent and aqueous mobile phases
- Rugged column stability
3 tilgængelige muligheder nedenfor
Product Details & Documents
The MAbPac™ HIC-10 column is based on high-purity, spherical, wide-pore silica particles functionalised with proprietary alkyl amide groups. Its advanced surface bonding technology leads to excellent chemical stability, high efficiency and unique selectivity optimised for MAbs and different from other HIC phases.
- Excellent biocompatibility and extremely low carryover
- High resolution and high efficiency
- Compatible with both organic solvent and aqueous mobile phases
- Rugged column stability
Specifications for Product Family
Specifikationer
- Catalog No:
- 555-7354
- 555-7355
- 555-7356
- Maks. tryk:
- 6000 psi
- 6000 psi
- 6000 psi
- pH-område:
- 2 - 8
- 2 - 8
- 2 - 8
- Temperature limit:
- 60 °C
- 60 °C
- 60 °C
Specifikationer
Product Family Options
Produktinformation
- Kolonne formPorestørrelsePartikelstørrelseØ indv.LængdePakkematerialeTilgængelighedPrice
- Analytisk1000 Å5 µm4,60 mm100 mmSpherical, fully porous ultrapure silicaLog ind for tilgængelighed1 * 1 Items11.110,00 krSpecifikationer:
More
Specifications
Cat. No.555-7354Maks. tryk6000 psipH-område2 - 8Temperature limit60 °C - Analytisk1000 Å5 µm4,60 mm250 mmSpherical, fully porous ultrapure silicaLog ind for tilgængelighed1 * 1 Items16.110,00 krSpecifikationer:
More
Specifications
Cat. No.555-7355Maks. tryk6000 psipH-område2 - 8Temperature limit60 °C
Guard columns
Produktinformation
- Kolonne formPorestørrelsePartikelstørrelseØ indv.LængdePakkematerialeTilgængelighedPrice
- Forkolonne1000 Å5 µm4,60 mm10 mmSpherical, fully porous ultrapure silicaLog ind for tilgængelighed1 * 2 Items1.900,00 krSpecifikationer:
More
Specifications
Cat. No.555-7356Maks. tryk6000 psipH-område2 - 8Temperature limit60 °C