Orders LinkContactUsLinkComponent
Forceps, micro, jeweller pattern, Aesculap®
 :   Aesculap
Forceps, micro, jeweller pattern, Aesculap®
 :  Aesculap
Forceps, micro, jeweller pattern, Aesculap®
Product Family Options
  • Model
    Længde
  • Angled tip, very fine (0,3 mm)
    110 mm
    1 * 1 Items
    1 * 1 Items - 348,00 kr
    348,00 kr
    1 * 1 Items
       348,00 kr
  • Straight, very fine (0,2 mm)
    110 mm
    1 * 1 Items
    1 * 1 Items - 229,00 kr
    229,00 kr
    1 * 1 Items
       229,00 kr
  • Straight, very fine (0,15 mm)
    110 mm
    1 * 1 Items
    1 * 1 Items - 265,00 kr
    265,00 kr
    1 * 1 Items
       265,00 kr
  • Curved tip, very fine (0,2 mm)
    115 mm
    1 * 1 Items
    1 * 1 Items - 306,00 kr
    306,00 kr
    1 * 1 Items
       306,00 kr
  • Straight, very fine (0,3 mm)
    135 mm
    1 * 1 Items
    1 * 1 Items - 322,00 kr
    322,00 kr
    1 * 1 Items
       322,00 kr
  • Curved tip, very fine (0,4 mm), with teeth 1:2
    115 mm
    1 * 1 Items
    1 * 1 Items
  • Straight, very fine (0,4 mm), with teeth 1:2
    135 mm