Long handle ESD stick, Stat-Rite®
Leverandør: Texwipe
Om denne vare
This ESD-safe stick with micro tips is a double-ended cleaning and prepping tool.
- Controls ESD without the need of carbons and ionic antistats
- Made from Stat-Rite®, an inherently dissipative polymer
Designed for use at ESD-safe workstations.
Advarsel: *The product(s) on this page are outside the scope of the EU Directive 2019/904.
2 tilgængelige muligheder nedenfor
Product Details & Documents
This ESD-safe stick with micro tips is a double-ended cleaning and prepping tool.
- Controls ESD without the need of carbons and ionic antistats
- Made from Stat-Rite®, an inherently dissipative polymer
Designed for use at ESD-safe workstations.
Advarsel: *The product(s) on this page are outside the scope of the EU Directive 2019/904.
Specifications for Product Family
Specifikationer
- Catalog No:
- TEXWTX769E
- 115-1901
- Hovedbredde:
- 1 mm
- 1 mm
- Håndtagslængde:
- 56,5 mm
- 56,5 mm
- Håndtagsbredde:
- 2,4 mm
- 2,4 mm
Specifikationer
Product Family Options
Produktinformation
- LængdeTilgængelighedPrice
- TEXWTX769EESD sticks with micro tipsSpecifikationer:New Product
More
Specifications
Cat. No.TEXWTX769EHovedbredde1 mmHåndtagslængde56,5 mmHåndtagsbredde2,4 mmNew Product - 115-1901ESD sticks with micro tipsSpecifikationer:
More
Specifications
Cat. No.115-1901Hovedbredde1 mmHåndtagslængde56,5 mmHåndtagsbredde2,4 mm