Orders LinkContactUsLinkComponent
Enzymatic digester, GDE
Enzymatic digester, GDE
Leverandør:   VELP SCIENTIFIC
Enzymatic digester, GDE
Leverandør:  VELP SCIENTIFIC
Certifikater
Enzymatic digester, GDE
1 tilgængelige muligheder nedenfor
Product Details & Documents
The procedure for determining dietary fibre exposes the sample to a series of enzymatic digestions that simulate the real digestive process which takes place in the human or animal digestive tract, calculating the undigested residue remaining at the end of the analysis. Generally dietary fibre analysis is carried out on foods intended for human consumption whereas raw fibre analysis is carried out on animal feeds or on raw materials of vegetable origin, eg. cereals. The GDE performs enzymatic digestion, a delicate phase where samples are immersed in a thermostatic water bath and stirred. Continuous and constant sample mixing is necessary in order to prevent the sample from overheating. The unit consists of an immersion thermostat, a transparent polycarbonate tank and a VELP 6-place magnetic stirrer to ensure excellent thermoregulation and precision. The use of deionised water is recommended as it eliminates any possible cause of limescale.
  • Intuitive digital display allows the required temperature to be set with accuracy of ±0,2 °C
  • Suitable for unattended operation
RT = Ambient
Specifications for Product Family
Specifikationer
  • Catalog No:
  • VELPSA30410209
  • VELPSA30400209
  • Temperaturinterval:
  • RT*...105 °C
  • RT*...105 °C
  • Varmekapacitet:
  • 900 W
  • 900 W
  • Ydermål B×D×H:
  • 413×410×295 mm
  • 413×410×295 mm
  • Vægt:
  • 6,2 kg
  • 6,2 kg
Specifikationer
Product Family Options
Produktinformation
  • Tilgængelighed
    Price
  • VELPSA30410209
    GDE enzymatic digester, 115 V, 50 - 60 Hz
    GDE enzymatic digester, 115 V, 50 - 60 Hz
    Discontinued
    Specifikationer:
    More
    Specifications
    Cat. No.
    VELPSA30410209
    Temperaturinterval
    RT*...105 °C
    Varmekapacitet
    900 W
    Ydermål B×D×H
    413×410×295 mm
    Vægt
    6,2 kg
  • VELPSA30400209
    GDE enzymatic digester, 230 V, 50 - 60 Hz
    GDE enzymatic digester, 230 V, 50 - 60 Hz
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 19.570,00 kr
    19.570,00 kr
    1 * 1 Items
       19.570,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    VELPSA30400209
    Temperaturinterval
    RT*...105 °C
    Varmekapacitet
    900 W
    Ydermål B×D×H
    413×410×295 mm
    Vægt
    6,2 kg