Self-laminating labels, MiniMark
Self-laminating labels, MNK-BPT-211-427, Vinyl, hvid/klar, 25×25 mm
Katalog # BRDY710113
Leverandør: Brady
Gå til:
Product Details & Documents
The MiniMark self-laminating wire and cable markers are designed to adhere to highly curved surfaces, wires and cables. They feature a white printable area with clear tail that wraps around marker. The clear tail protects the text, providing superior resistance to abrasion, dirt, oils, solvents, water and excessive handling.
Specifikationer
- Farve:hvid/klar
- Beskrivelse:Self-laminating labels
- Dimensioner:25×25 mm
- Materiale:Vinyl
- Model:MNK-BPT-211-427