Orders LinkContactUsLinkComponent
Cleanroom labels, matte
Cleanroom labels, matte
Leverandør:   Texwipe
Cleanroom labels, matte
Leverandør:  Texwipe
Certifikater
Cleanroom labels, matte
Cleanroom labels, matte
3 tilgængelige muligheder nedenfor
Product Details & Documents
Matte finish cleanroom labels provide excellent compatibility with cleanroom pens and thermal transfer inks. Available with two different adhesive levels to provide excellent adhesion to a variety of surfaces without leaving residue.
  • Ideal for handwritten applications
Product Family Options
Produktinformation
  • Materiale
    Klæbestof
    Farve
    Dimensioner
    Pakket
    Tilgængelighed
    Price
  • 817-0301
    Removable cleanroom labels, matte finish
    Removable cleanroom labels, matte finish
    PP
    Akryl
    hvid
    38×38 mm
    1000 labels/roll
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 4 ROL
    1 * 4 ROL - 4.790,00 kr
    4.790,00 kr
    1 * 4 ROL
       4.790,00 kr
  • 817-0305
    Removable cleanroom labels, matte finish
    Removable cleanroom labels, matte finish
    PP
    Akryl
    hvid
    76×51 mm
    1000 labels/roll
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 4 ROL
    1 * 4 ROL - 6.270,00 kr
    6.270,00 kr
    1 * 4 ROL
       6.270,00 kr
  • 817-0308
    Removable cleanroom labels, matte finish
    Removable cleanroom labels, matte finish
    PP
    Akryl
    hvid
    102×51 mm
    1000 labels/roll
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 4 ROL
    1 * 4 ROL - 6.280,00 kr
    6.280,00 kr
    1 * 4 ROL
       6.280,00 kr