Orders LinkContactUsLinkComponent
Accessories for face shields
Accessories for face shields
Ergonomic headband with foam sweatband and 90° flip-up function, protects against electric arcs, blue, for head sizes 53 - 61 cm
Katalog # UNVT605.04.00.00
Leverandør:   UNIVET
Accessories for face shields
Katalog # UNVT605.04.00.00
Leverandør:  UNIVET
Accessories for face shields
Accessories for face shields
Ergonomic headband with foam sweatband and 90° flip-up function, protects against electric arcs, blue, for head sizes 53 - 61 cm
Price
201,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This ergonomic headgear is made from anti-allergic material. With upper band for height adjustment of the cap.
  • Interchangeable front foam sweatband
  • Adjustment of the head circumference by rack
  • 90° flip-up through friction
  • Free of exposed metal parts for protection against electric arcs
Specifikationer
  • Beskrivelse:
    Ergonomic headband with foam sweatband and 90° flip-up function, protects against electric arcs, blue, for head sizes 53 - 61 cm