Multi-layer adhesive floor mats, Nomad™ 4300
Catalog # 115-2706
Supplier: 3M
Specifications
- Farve:hvid
- L×W:600×1150 mm
- Seq:07
- Environmentally Preferable:
- Pakn:6
Specifications
About this item
4300 contamination control mats consist of 40 layers of adhesive-coated clear polyethylene sheets attached to a vinyl backing. For use at cleanroom entrances, hospitals, semi-conductor, medical device, pharmaceutical, biological and nuclear industries, also at factory to office entrances and other work areas requiring high levels of cleanliness.
- Backing adheres to the floor, while the tacky top surface removes dirt and contaminants from the bottom of shoes and wheels
- Very flat; no frame needed
Colour: Blue or white