Slide mailer
Slide mailers, PP, ⁵/₁₀
Katalog # LPIT290400
Leverandør: LP ITALIANA
Gå til:
Product Details & Documents
Slide mailer, ideal for storing, shipping and staining.
- Dimensions: Ø 48×86
Specifikationer
- Antal objektglas:⁵/₁₀
- Beskrivelse:Slide mailers
- Materiale:PP
- Seq:01