Casting dams, Owl™
Supplier: THERMO OWL SCIENTIFIC
About this item
3 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
- Catalog No:
- OWLADAM-13
- OWLADAM-20
- OWLADAM-23
- Model:
- DAM-13
- DAM-20
- DAM-23
- Gelstørrelse B×L:
- 130×130 mm
- 200×200 mm
- 230×230 mm
Specifications
Product Family Options
Product Information
- BeskrivelseModelAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.OWLADAM-13ModelDAM-13Gelstørrelse B×L130×130 mm - Specifications:
More
Specifications
Cat. No.OWLADAM-20ModelDAM-20Gelstørrelse B×L200×200 mm - Specifications:
More
Specifications
Cat. No.OWLADAM-23ModelDAM-23Gelstørrelse B×L230×230 mm