ContactUsLinkComponent
Combs for horizontal gel electrophoresis system, Owl™ D4
  730-0231
undefined
Combs for horizontal gel electrophoresis system, Owl™ D4
  730-0231
  • Beskrivelse:
    Comb, micro-well 17- tooth,1,5 mm thick
  • Seq:
    02
  • Pakn:
    1