Orders LinkContactUsLinkComponent
Microplate stands
Microplate stands
Supplier:   DIVERSIFIED BIOTECH
Microplate stands
Certificates
Microplate stands
2 Options Available Below
Product Details & Documents
Convenient holder for microplates to comfortably view samples and pipette without strain. Available tilted at a 30° angle or as an adjustable version allowing the flexibility to adjust the angle of the microplate (45° max. tilt).
Product Family Options
Product Information
  • Beskrivelse
    Availability
    Price
  • Well plate stand, adjustable up to 45° angle
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.000,00 kr
    2.000,00 kr
    1 * 1 Items
       2.000,00 kr
  • Well plate stand, tilted at 30° angle
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 967,00 kr
    967,00 kr
    1 * 1 Items
       967,00 kr