Scoops, Rotilabo®
Scoop set, incl. 7 scoops with capacity from 25 - 1520 ml, 25 - 1520 ml, 135 - 400 mm, 42 - 170 mm, HDPE, hvid
Catalog # ROTH0997.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Set of plastic scoops made of white HDPE.
Caution: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Specifications
- Bredde:42 - 170 mm
- Kapacitet:25 - 1520 ml
- Farve:hvid
- Beskrivelse:Scoop set, incl. 7 scoops with capacity from 25 - 1520 ml
- Længde:135 - 400 mm
- Materiale:HDPE
- Seq:08