Hand towels
Paper towel rolls, embossed, 1 ply, grå, 190 m × 200 mm, 6 rolls à 190 m
Katalog # KIMW6063
Leverandør: KIMBERLY CLARK
Gå til:
Product Details & Documents
Embossed paper towel rolls for use with CONTROLOMATIC* dispenser 6963.
Documents
KIMB6063_TDS_EN
Datablad
Specifikationer
- Pakket:6 rolls à 190 m
- Farve:grå
- Beskrivelse:Paper towel rolls, embossed, 1 ply
- L×B:190 m × 200 mm