Softshell jackets for ladies and men, Acode® Advanced
Leverandør: FRISTADS KANSAS
Om denne vare
Lightweight 4-way softshell jacket made of 90% polyester and 10% elasthane with a high level of breathability. Available for ladies and for men. The version for ladies features a tailored fit.
- Water-repellent
- Red details and zip pullers
- Front pockets with zip, breast pocket with zip
- Adjustable cuffs and drawstring at hem
- Extended back
7 tilgængelige muligheder nedenfor
Product Details & Documents
Lightweight 4-way softshell jacket made of 90% polyester and 10% elasthane with a high level of breathability. Available for ladies and for men. The version for ladies features a tailored fit.
- Water-repellent
- Red details and zip pullers
- Front pockets with zip, breast pocket with zip
- Adjustable cuffs and drawstring at hem
- Extended back
Specifications for Product Family
Specifikationer
- Catalog No:
- KWIN110173940S
- KWIN110173940M
- KWIN110173940L
- KWIN110172940S
- KWIN110172940XL
- KWIN1101729402XL
- KWIN1101729403XL
- Outfit applikationer:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Køn:
- For kvinder
- For kvinder
- For kvinder
- For mænd
- For mænd
- For mænd
- For mænd
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Lukkestørrelse:
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Stof:
- 90% polyester/10% elasthane
- 90% polyester/10% elasthane
- 90% polyester/10% elasthane
- 90% polyester/10% elasthane
- 90% polyester/10% elasthane
- 90% polyester/10% elasthane
- 90% polyester/10% elasthane
Specifikationer
Product Family Options
For ladies
Produktinformation
- FarveTøj størrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.KWIN110173940SOutfit applikationerOutdoorRisk areaRain (waterproof)KønFor kvinderSleeve or leg lengthLongLukkestørrelseMed lynlåsStof90% polyester/10% elasthane - Specifikationer:
More
Specifications
Cat. No.KWIN110173940MOutfit applikationerOutdoorRisk areaRain (waterproof)KønFor kvinderSleeve or leg lengthLongLukkestørrelseMed lynlåsStof90% polyester/10% elasthane - Specifikationer:
More
Specifications
Cat. No.KWIN110173940LOutfit applikationerOutdoorRisk areaRain (waterproof)KønFor kvinderSleeve or leg lengthLongLukkestørrelseMed lynlåsStof90% polyester/10% elasthane
For men
Produktinformation
- FarveTøj størrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.KWIN110172940SOutfit applikationerOutdoorRisk areaRain (waterproof)KønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof90% polyester/10% elasthane - Specifikationer:
More
Specifications
Cat. No.KWIN110172940XLOutfit applikationerOutdoorRisk areaRain (waterproof)KønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof90% polyester/10% elasthane - Specifikationer:
More
Specifications
Cat. No.KWIN1101729402XLOutfit applikationerOutdoorRisk areaRain (waterproof)KønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof90% polyester/10% elasthane - Specifikationer:
More
Specifications
Cat. No.KWIN1101729403XLOutfit applikationerOutdoorRisk areaRain (waterproof)KønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof90% polyester/10% elasthane