Orders LinkContactUsLinkComponent
Micro Tubes with Thread
Micro Tubes with Thread
Marketsource
Supplier:   SARSTEDT
Micro Tubes with Thread
Supplier:  SARSTEDT
Marketsource
Certificates
Micro Tubes with Thread
2 Options Available Below
Product Details & Documents
PP, translucent with PP screw cap.
  • With printed label
  • Withstand centrifugation up to 20000×g
  • Attached, not screwed-on cap
  • Sterile and free of DNA, DNase/RNase, PCR inhibitors, ATP and pyrogens/endotoxins
With or without skirted base.
Product Family Options
Tubes, 0,5 ml, with skirted base, neutral screw cap
Product Information
  • Beskrivelse
    Kapacitet
    Pakket
    Availability
    Price
  • Micro tubes with skirted base, neutral cap
    0,5 ml
    500/case
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items
    Specifications:
    Marketsource
    Marketsource
Tubes, 2 ml, without skirted base, neutral screw cap
Product Information
  • Beskrivelse
    Kapacitet
    Pakket
    Availability
    Price
  • Micro tubes without skirted base, neutral cap
    2,0 ml
    1000/case
    Log in for availability
    Log in for availability
    1 * 1.000 Items
    1 * 1.000 Items
    Specifications:
    Marketsource
    Marketsource