Kidney dishes
260×100×55 mm
Katalog # SEMA1254
Leverandør: SEMADENI
Gå til:
Product Details & Documents
Kidney dishes made of white PP.
- Temperature resistant up to 130 °C
- Excellent chemical resistance
Specifikationer
- B×D×H:260×100×55 mm