Sharps disposal container Multi-Safe steri
Katalog # SARS77.3899.060
Leverandør: SARSTEDT
Specifikationer
- Kapacitet:60,0 l
- Beskrivelse:steri 60
- B×D×H:393x393x520 mm
- Pakn:120
Specifikationer
Om denne vare
Box, made of PP, yellow/red. Secure and robust container for the disposal of infectious wastes that can be deactivated by autoclaving at 121 or 134 °C.
- The small cover melts at about 90°C to enable autoclaving. A grate prevents waste from falling out
- The box is stackable both empty and filled
- Robust handle in the lid and two moulded recesses in the box