Orders LinkContactUsLinkComponent
Safety boxes Multi-Safe vario
Safety boxes Multi-Safe vario
vario 1500, 1,5 l, gul/rød, 244x104x20 mm
Katalog # SARS77.3893.015
Marketsource
Leverandør:   SARSTEDT
Safety boxes Multi-Safe vario
Katalog # SARS77.3893.015
Leverandør:  SARSTEDT
Marketsource
Safety boxes Multi-Safe vario
Safety boxes Multi-Safe vario
vario 1500, 1,5 l, gul/rød, 244x104x20 mm
Price
Spørg for pris
Log ind for tilgængelighed
1 * 60 Items
Product Details & Documents
Boxes, made of PP, yellow/red. With the large deposit slot, the Multi-Safe vario is suitable for the disposal of all small bulky wastes.
  • Suitable for use on a syringe tray and Safety-Tray
  • Supplied with a carrying handle and still stackable to save space
 : TRBA 250
Specifikationer
  • Kapacitet:
    1,5 l
  • Farve:
    gul/rød
  • Beskrivelse:
    vario 1500
  • Seq:
    01
  • B×D×H:
    244x104x20 mm