Multi-Safe sani 200 Secure Sharp Container
sani 200, 200 ml, 70x131 mm
Catalog # SARS77.3890.002
Marketsource
Supplier: SARSTEDT
Jump to:
Product Details & Documents
Boxes, made of PP, yellow/red. Secure sharp container with final closure in a handy size for doctors, nurses, rescue services- and ambulatory medical services, or for the doctors and rescue services supply bag.
- Single-handed opening and closing of day lid
- U-Shaped recess for sharps removal from Luer lock systems
Certifications: TRBA 250, UN, DIN EN ISO 23907
Specifications
- Diameter×H:70x131 mm
- Kapacitet:200 ml
- Beskrivelse:sani 200