Orders LinkContactUsLinkComponent
Immersion flow cups, 'Ford'
Immersion flow cups, 'Ford'
Viscosity cup, "Ford", 70 - 370 cSt, 4,1 mm
Catalog # SIMEVF2087
Supplier:   Industrial Physics
Immersion flow cups, 'Ford'
Catalog # SIMEVF2087
Supplier:  Industrial Physics
Immersion flow cups, 'Ford'
Immersion flow cups, 'Ford'
Viscosity cup, "Ford", 70 - 370 cSt, 4,1 mm
Price
1.640,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Titanium anodised cup with a fixed stainless steel nozzle and handle.
Certifications: ASTM D1200
Delivery information: Supplied with a hard plastic case.
Documents
Specifications
  • Område:
    70 - 370 cSt
  • Beskrivelse:
    Viscosity cup, "Ford"
  • Bægeråbning:
    4,1 mm
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • SIMEDI0076
    Digital stopwatch, 0,01 s
    Log in for availability
    1 * 1 Items - 221,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • SIMEVF2062
    Ring stand fro flow cups
    Log in for availability
    1 * 1 Items - 625,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 769-1151
    Viscosity convertor of flow time to viscosity
    Log in for availability
    1 * 1 Items - 653,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 769-1131
    Temperature control jacket with tripod for DIN and ASTM cups
    Log in for availability
    1 * 1 Items - 4.290,00 kr