Orders LinkContactUsLinkComponent
Microscope Slide Boxes
Microscope Slide Boxes
Katalog #
Microscope Slide Boxes
Katalog #
Leverandør:  
Microscope Slide Boxes
Microscope Slide Boxes
Product Details & Documents
Heavy-duty, two-piece microscope slide boxes are ideal for storing or transportating standard slides, 75×25 mm (3×1").
  • Designed to securely store slides in grooved slots
  • Molded feet secure stacking for easy transport and storage
Closes securely with a rust-resistant nickel-plated clasp and pin hinges. ABS plastic boxes available with either cork or foam lining.
Documents