Analytical funnels
10 mm, 80 mm
Katalog # DELT19150
Leverandør: DELTALAB
Gå til:
Product Details & Documents
PP, translucent.
- With long stem and grooves
- Rigidified by a thickened edge
- Rapid flow due to a steep 60° angle
Designed for use with standard filter paper.
Specifikationer
- Tragt udv. Ø:80 mm
- Stilk udv. Ø:10 mm