Orders LinkContactUsLinkComponent
Basic ESD Chairs
Basic ESD Chairs
Leverandør:   bimos
Basic ESD Chairs
Leverandør:  bimos
Certifikater
Basic ESD Chairs
Basic ESD Chairs
6 tilgængelige muligheder nedenfor
Product Details & Documents
An ergonomic design, superb cleanroom properties and reliable ESD measures are a lot to ask from a workplace chair. Yet Cleanroom Basic ESD meets all these criteria and offers a high level of quality in the process.
  • Conductive high-comfort upholstery: choice of fabric or synthetic leather
  • Seat and backrest height adjustment
  • The frame is available in either black or light grey
  • Choice of three different mechanisms: contact backrest, contact backrest with seat tilt adjustment and synchronous mechanism with weight regulation
  • Steel components with conductive coating
  • Five volume conductive castors/glides
  • With five star base
Basic ESD has conductive covers. It features a sturdy five-legged base frame with flat lines, which is made from epoxyresin coated sectional steel tubing and provides good stability.
Certifications: Conform to DIN 68877, discharge and surface resistance according to EN 61340-5-1
German GS 'safety tested' certificate
Quality Management System DIN EN ISO 9001
Regulation EWG Nr. 1836/93 and
Environmental Management System DIN EN ISO 14001
Product Family Options
Produktinformation
  • Højde
    Størrelse
    med
    Tilgængelighed
    Price
  • BIMO9151E-2571
    Basic ESD 2 chair, made from skai® synthetic leather, black, with contact backrest, backrest height: 430 mm
    Basic ESD 2 chair, made from skai® synthetic leather, black, with contact backrest, backrest height: 430 mm
    470 - 610 mm
    Castors
    Five-legged base
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr4,450.00
    kr4,450.00
    1 * 1 Items
       kr4,450.00
  • BIMO9158E-2571-521
    Basic ESD 2 chair, made from Duotec ESD fabric, black, with contact backrest with tilting seat, backrest height: 490 mm
    Basic ESD 2 chair, made from Duotec ESD fabric, black, with contact backrest with tilting seat, backrest height: 490 mm
    490 - 630 mm
    Castors
    Five-legged base
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr5,480.00
    kr5,480.00
    1 * 1 Items
       kr5,480.00
  • BIMO9155E-238-9801
    Basic ESD 2 chair, made from Duotec ESD fabric, black, with contact backrest with tilting seat, backrest height: 530 mm
    Basic ESD 2 chair, made from Duotec ESD fabric, black, with contact backrest with tilting seat, backrest height: 530 mm
    470 - 610 mm
    Castors
    Five-legged base
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr5,020.00
    kr5,020.00
    1 * 1 Items
       kr5,020.00
  • BIMO9155E257123852
    Basic ESD 2 chair, made from Skai® ESD synthetic leather, with contact backrest with tilting seat, backrest height: 530 mm
    Basic ESD 2 chair, made from Skai® ESD synthetic leather, with contact backrest with tilting seat, backrest height: 530 mm
    470 - 610 mm
    Castors
    Five-legged base
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr5,280.00
    kr5,280.00
    1 * 1 Items
       kr5,280.00
  • BIMO9155E-9802
    Basic ESD 2 chair, made from Duotec ESD fabric, blue, with contact backrest with tilting seat, backrest height: 530 mm
    Basic ESD 2 chair, made from Duotec ESD fabric, blue, with contact backrest with tilting seat, backrest height: 530 mm
    470 - 610 mm
    Castors
    Five-legged base
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr5,020.00
    kr5,020.00
    1 * 1 Items
       kr5,020.00
  • BIMO9158E-9801
    Basic ESD 2 chair, made from Duotec ESD fabric, black, with synchronous mechanism with weight regulation, backrest height: 530 mm
    Basic ESD 2 chair, made from Duotec ESD fabric, black, with synchronous mechanism with weight regulation, backrest height: 530 mm
    470 - 610 mm
    Castors
    Five-legged base
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr5,390.00
    kr5,390.00
    1 * 1 Items
       kr5,390.00