Gel loading pipette tips, MµltiFlex®
Flat, 0,2 mm, 0,5 - 200 µl, -, Transparent, 200/rack
Katalog # 612-0675
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
These tips are suitable for loading SDS page protein gels, horizontal agarose and DNA sequencing gels.
- Free of RNAse/DNAse and pyrogens
- Free of human DNA and PCR inhibitors
- Autoclavable
Suitable for Brand, Eppendorf, Finnpipette®, Gilson, Oxford, Roth, Sartorius, Socorex.
Specifikationer
- Pakket:200/rack
- Steril:-
- Volumen:0,5 - 200 µl
- Spidstype:Flat, 0,2 mm
- Spits farge:Transparent