Cover spectacles, Visitor
Anti-UV, transparent, Clear PC (2C-1.2 3M 1 F)
111-1554
: 3M
These basic cover glasses (OTG) with clear PC lens and a transparent frame fit securely and comfortably over any prescription spectacles, but can also be worn alone. They offer good side and brow protection and are very light.
- Straight, flat and length-adjustable temples offer excellent compatibility with earmuffs
- Can be worn over prescription spectacles
- Four-stage adjustable temple length and adjustable lens angle for optimum fit
- Soft, narrow temple tips
Ideal as classic visitor spectacles, for example in the construction industry or in forestry.
: Compliant with EN 166, EN 170; 3M EN 166 F CE (frame)
- Model:Anti-UV
- Farve:transparent
- Linser:Clear PC (2C-1.2 3M 1 F)