Urine collection cups, disposable
DELT309324
: DELTALAB
- Beskrivelse:Urine container set with green cap
- Seq:01
- Pakn:300
Urine collection system designed to save space in the lab, and to be used when urine samples need to be collected into tubes for a subsequent centrifugation.
- Manufactured under aseptic conditions
The patient urinates in the 150 ml plastic cup and then pours the urine into the tube. Once the tube is filled up, the patient caps it and delivers the sample to the lab, then label filled with the required data. The patient just has to follow the hospital or laboratory instructions.
: Includes bag printed with instruction for use, cup PS (150 ml), tube PP (10 ml), cap and label.
: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904