Cut resistant gloves, Sharpflex Latex
Om denne vare
These gloves knitted from composite and para-aramid yarn features a blue crinkled latex coating on the palm.
- Knitting technique provides the glove with excellent fit and high dexterity
- Para-aramid yarn protects against contact heat and cuts
- Plated polyamide inside: Limit the contact of the skin with composite fibre
- Latex coating provides better grip and light thermal protection (up to 250 °C)
Ideal for handling heavy parts with high cut risks in humid and slippery environments. The gloves also provide light thermal protection (up to 250 °C).
Certifications: EN 388:2016 (2X42C), EN 407 (X2XXXX); Cat. II
3 tilgængelige muligheder nedenfor
Product Details & Documents
These gloves knitted from composite and para-aramid yarn features a blue crinkled latex coating on the palm.
- Knitting technique provides the glove with excellent fit and high dexterity
- Para-aramid yarn protects against contact heat and cuts
- Plated polyamide inside: Limit the contact of the skin with composite fibre
- Latex coating provides better grip and light thermal protection (up to 250 °C)
Ideal for handling heavy parts with high cut risks in humid and slippery environments. The gloves also provide light thermal protection (up to 250 °C).
Certifications: EN 388:2016 (2X42C), EN 407 (X2XXXX); Cat. II
Specifications for Product Family
Specifikationer
- Catalog No:
- DALL2232525/7
- DALL2232525/9
- DALL2232525/10
- Model:
- Latex
- Latex
- Latex
- Ambidextrous/Hånd-specifik:
- Hånd-specifik
- Hånd-specifik
- Hånd-specifik
- Engangs/Genbrugelig:
- Genanvendelig
- Genanvendelig
- Genanvendelig
- Foringsmateriale:
- Para-aramid
- Para-aramid
- Para-aramid
- Overflade:
- Crinkled coating on palm and fingertips
- Crinkled coating on palm and fingertips
- Crinkled coating on palm and fingertips
- Manchet:
- Strikket
- Strikket
- Strikket
- EN norm:
- EN 388:2016 (2X42C), EN 407 (X2XXXX)
- EN 388:2016 (2X42C), EN 407 (X2XXXX)
- EN 388:2016 (2X42C), EN 407 (X2XXXX)
Specifikationer
Product Family Options
Produktinformation
- Handske størrelseFarveLængdeTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.DALL2232525/7ModelLatexAmbidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialePara-aramidOverfladeCrinkled coating on palm and fingertipsManchetStrikketEN normEN 388:2016 (2X42C), EN 407 (X2XXXX) - Specifikationer:
More
Specifications
Cat. No.DALL2232525/9ModelLatexAmbidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialePara-aramidOverfladeCrinkled coating on palm and fingertipsManchetStrikketEN normEN 388:2016 (2X42C), EN 407 (X2XXXX) - Specifikationer:
More
Specifications
Cat. No.DALL2232525/10ModelLatexAmbidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialePara-aramidOverfladeCrinkled coating on palm and fingertipsManchetStrikketEN normEN 388:2016 (2X42C), EN 407 (X2XXXX)