Cut resistant gloves, Sharpflex Nit
Om denne vare
These gloves knitted from composite and para-aramid yarn features a black nitrile foam coating on the palm.
- Knitting technique provides the glove with excellent fit and high dexterity
- Para-aramid yarn protects against contact heat and cuts
- Plated polyamide inside: Limit the contact of the skin with composite fiber
- Nitrile coating provides good protection against oil and grease
- Foam coating is more flexible than standard nitrile coating
Ideal for handling heavy parts with high cut risks in oily and greasy environments.
Certifications: EN 388:2016 (3X42C), EN 407 (X1XXXX); Cat. II
4 tilgængelige muligheder nedenfor
Product Details & Documents
These gloves knitted from composite and para-aramid yarn features a black nitrile foam coating on the palm.
- Knitting technique provides the glove with excellent fit and high dexterity
- Para-aramid yarn protects against contact heat and cuts
- Plated polyamide inside: Limit the contact of the skin with composite fiber
- Nitrile coating provides good protection against oil and grease
- Foam coating is more flexible than standard nitrile coating
Ideal for handling heavy parts with high cut risks in oily and greasy environments.
Certifications: EN 388:2016 (3X42C), EN 407 (X1XXXX); Cat. II
Specifications for Product Family
Specifikationer
- Catalog No:
- DALL2232524/7
- DALL2232524/9
- DALL2232524/10
- DALL2232524/11
- Model:
- Nit
- Nit
- Nit
- Nit
- Ambidextrous/Hånd-specifik:
- Hånd-specifik
- Hånd-specifik
- Hånd-specifik
- Hånd-specifik
- Engangs/Genbrugelig:
- Genanvendelig
- Genanvendelig
- Genanvendelig
- Genanvendelig
- Foringsmateriale:
- Para-aramid
- Para-aramid
- Para-aramid
- Para-aramid
- Overflade:
- Foam coating on palm and fingertips
- Foam coating on palm and fingertips
- Foam coating on palm and fingertips
- Foam coating on palm and fingertips
- Måler:
- 13
- 13
- 13
- 13
- Manchet:
- Strikket
- Strikket
- Strikket
- Strikket
- EN norm:
- EN 388:2016 (3X42C), EN 407 (X1XXXX)
- EN 388:2016 (3X42C), EN 407 (X1XXXX)
- EN 388:2016 (3X42C), EN 407 (X1XXXX)
- EN 388:2016 (3X42C), EN 407 (X1XXXX)
Specifikationer
Product Family Options
Produktinformation
- Handske størrelseFarveLængdeAvailabilityPrice
- Specifikationer:
More
Specifications
Cat. No.DALL2232524/7ModelNitAmbidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialePara-aramidOverfladeFoam coating on palm and fingertipsMåler13ManchetStrikketEN normEN 388:2016 (3X42C), EN 407 (X1XXXX) - Specifikationer:
More
Specifications
Cat. No.DALL2232524/9ModelNitAmbidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialePara-aramidOverfladeFoam coating on palm and fingertipsMåler13ManchetStrikketEN normEN 388:2016 (3X42C), EN 407 (X1XXXX) - Specifikationer:
More
Specifications
Cat. No.DALL2232524/10ModelNitAmbidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialePara-aramidOverfladeFoam coating on palm and fingertipsMåler13ManchetStrikketEN normEN 388:2016 (3X42C), EN 407 (X1XXXX) - Specifikationer:
More
Specifications
Cat. No.DALL2232524/11ModelNitAmbidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialePara-aramidOverfladeFoam coating on palm and fingertipsMåler13ManchetStrikketEN normEN 388:2016 (3X42C), EN 407 (X1XXXX)