Orders LinkContactUsLinkComponent
Cell detachment solution, Accutase
Cell detachment solution, Accutase
Accutase, 100 ml
Catalog # L0950-100
Supplier:   Biowest
Cell detachment solution, Accutase
Catalog # L0950-100
Supplier:  Biowest
CAS Number:  
Cell detachment solution, Accutase
Cell detachment solution, Accutase
Accutase, 100 ml
CAS Number:  
Price
1.270,00 kr
Log in for availability
1 * 100 mL
Product Details & Documents
Accustase is a ready to use cell detachment solution of proteolytic and collagenolytic enzymes. Accutase is a direct gentle replacement for trypsin solution. It can also be used on suspension cells to reduce their agglutination in order to ease counting.
Documents
Specifications
Specifications
  • Pakn:
    100 ml
  • Beskrivelse:
    Accutase