Labelling tapes, TC291
Laminated tape, TC291, Polyester, sort/hvid, 9 mm×8 m
Katalog # BROTTC-291
Leverandør: BROTHER
Gå til:
Product Details & Documents
Laminated black on white polyester tape.
Length: 8,0 m
Specifikationer
- Model:TC291
- Farve:sort/hvid
- Beskrivelse:Laminated tape
- Dimensioner:9 mm×8 m
- Materiale:Polyester