Flasks, round bottom, with three parallel necks
100 ml, 14/23 NS, 14/23 NS
Katalog # NEUB0760-0100-14
Leverandør: NEUBERT VOLUME GLASSWAERE
Flasks, round bottom, with three parallel necks
Katalog # NEUB0760-0100-14
Leverandør: NEUBERT VOLUME GLASSWAERE
Gå til:
Product Details & Documents
Borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
Specifikationer
- Kapacitet:100 ml
- Seq:02
- Sidehalse:14/23 NS
- Centreret hals:14/23 NS