Orders LinkContactUsLinkComponent
Accessories standard pH electrodes
Adapter for the plug-in of electrodes with BNC-plugs to devices with Cinch-sockets
  GRSG601382
 :   Greisinger
Accessories standard pH electrodes
  GRSG601382
Accessories standard pH electrodes
Accessories standard pH electrodes
Adapter for the plug-in of electrodes with BNC-plugs to devices with Cinch-sockets
1 * 1 Items
  • Beskrivelse:
    Adapter for the plug-in of electrodes with BNC-plugs to devices with Cinch-sockets