Heating Cords
Leverandør: WINKLER
Gå til:
Om denne vare
Heating cord which consists of one heating spiral insulated with glass-silk.
- With flexible glass-silk outer braiding
- Max. temperature 400 °C
9 tilgængelige muligheder nedenfor
Product Details & Documents
Heating cord which consists of one heating spiral insulated with glass-silk.
- With flexible glass-silk outer braiding
- Max. temperature 400 °C
Product Family Options
Produktinformation
- LængdeTilgængelighedPrice
- WINKWKG00400500075Heating cord, 75 W
- WINKWKG00401000150Heating cord, 150 W
- WINKWKG00401500225Heating cord, 225 W
- WINKWKG00402500375Heating cord, 375 W
- WINKWKG00401000300Heating cord, 300 W
- WINKWKG00403000450Heating cord, 450 W
- WINKWKG00404000600Heating cord, 600 W
- WINKWKG00405000750Heating cord, 750 W
- WINKWKG00406000900Heating cord, 900 W