Orders LinkContactUsLinkComponent
Storage bottles, rectangular, PE, obong, without caps
Storage bottles, rectangular, PE, obong, without caps
118 ml
Katalog # HACH2318406
Leverandør:   Hach
Storage bottles, rectangular, PE, obong, without caps
Katalog # HACH2318406
Leverandør:  Hach
Storage bottles, rectangular, PE, obong, without caps
Storage bottles, rectangular, PE, obong, without caps
118 ml
Price
kr124.00
Log ind for tilgængelighed
1 * 6 Items
Product Details & Documents
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
Economical and robust bottles for storing liquids.
Specifikationer
  • Kapacitet:
    118 ml
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Dropper cap, with 0,5 and 1,0 ml marks on glass barrel
    Dropper cap, with 0,5 and 1,0 ml marks on glass barrel
    HACH2318506
    Log ind for tilgængelighed
    1 * 6 Items - 221,00 kr