Sample Bags
Sampling bags, sodium thiosulphate pill (10 mg), 100 ml
Katalog # HACH2075333
Leverandør: Hach
Gå til:
Product Details & Documents
They are transparent PE bags that are specially designed for chlorinated water samples and also for testing recreational water, waste water and drinking water.
- Sterile and disposable bags
- With 'Whirl-Pak' closure
- Write-on strip for waterproof ink
Certifications: Complies with USEPA.
Specifikationer
- Kapacitet:100 ml
- Beskrivelse:Sampling bags, sodium thiosulphate pill (10 mg)