Transparent classic human skull model
Transparent classic human skull model, in three parts, 0,6 kg, 200×135×155 mm
Katalog # BINH1000051
Marketsource
Leverandør: 3B Scientific
Gå til:
Product Details & Documents
Use this unique transparent replica of the human skull to study internal structures that otherwise are visible only through x-ray images.
- High-quality original casts
- Skull is handmade of hard, unbreakable plastic
- Highly accurate representation of the fissures, foramina, processes, sutures etc.
- Can be disassembled into Skull Cap, Base of Skull and Mandible
- As an option, you can insert a five-part brain (C 18) into the skull
Specifikationer
- Vægt:0,6 kg
- Beskrivelse:Transparent classic human skull model, in three parts
- B×D×H:200×135×155 mm