Screw caps, BOLA
S 40, koksgrå
Katalog # BOHLH988-40
Leverandør: Bohlender
Gå til:
Product Details & Documents
Screw caps, made of PPS, with handy knurl, with integrated PTFE/silicone gasket for compensation of unevenness on the sealing surface.
- Universal chemical resistance
- Temperature resistance: from –20 to +230 °C
After assembly, the product is only exposed to PTFE. The cap has high mechanical and thermal (up to +230 °C) capacitites.
Specifikationer
- Gevind:S 40
- Farve:koksgrå