Orders LinkContactUsLinkComponent
Silicone foam stoppers, conical
Silicone foam stoppers, conical
Certificates
Silicone foam stoppers, conical
Silicone foam stoppers, conical
11 Options Available Below
Product Details & Documents
These coloured conical stoppers are made from silicone foam.
  • Feather-light
  • Neutral odour
  • Physiologically inert
  • Temperature-resistant from –60 to +200 °C
Product Family Options
Product Information
  • Farve
    Ant. huller
    Højde
    Ø bund
    Ø top
    Availability
    Price
  • hvid
    0
    20,0 mm
    8,0 mm
    12,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 35,70 kr
    35,70 kr
    1 * 1 Items
       35,70 kr
  • lyserød
    0
    22,0 mm
    11,0 mm
    15,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 35,50 kr
    35,50 kr
    1 * 1 Items
       35,50 kr
  • gul
    0
    25,0 mm
    13,0 mm
    17,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 38,70 kr
    38,70 kr
    1 * 1 Items
       38,70 kr
  • lysegrøn
    0
    27,0 mm
    15,5 mm
    20,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 151,00 kr
    151,00 kr
    1 * 1 Items
       151,00 kr
  • blå
    0
    30,0 mm
    19,0 mm
    25,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 70,80 kr
    70,80 kr
    1 * 1 Items
       70,80 kr
  • hvid
    0
    33,0 mm
    23,0 mm
    30,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 89,60 kr
    89,60 kr
    1 * 1 Items
       89,60 kr
  • lyserød
    0
    35,0 mm
    28,0 mm
    35,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 138,00 kr
    138,00 kr
    1 * 1 Items
       138,00 kr
  • gul
    0
    40,0 mm
    33,0 mm
    40,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 172,00 kr
    172,00 kr
    1 * 1 Items
       172,00 kr
  • grøn
    0
    42,0 mm
    38,0 mm
    46,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 272,00 kr
    272,00 kr
    1 * 1 Items
       272,00 kr
  • blå
    0
    48,0 mm
    44,0 mm
    52,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 403,00 kr
    403,00 kr
    1 * 1 Items
       403,00 kr
  • hvid
    0
    52,0 mm
    49,0 mm
    58,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 445,00 kr
    445,00 kr
    1 * 1 Items
       445,00 kr