Vand for TOC, Supelco®
Glass bottle, 1 L
Leverandør: Merck
Vand for TOC, Supelco®
Katalog # 78533-1L
Leverandør: Merck
CAS nummer: 7732-18-5
Gå til:
Product Details & Documents
Synonymer: Hydrogen oxide
- CAS nummer:7732-18-5
- Formel:H₂O
- MDL nummer:MFCD00011332
- Molecular Weight:18.02 g/mol
- Merck Index:13,10098
- Kogende Pt:100 °C (1013 hPa)
- Smeltende Pt:0 °C
- Tæthed:1 g/cm³ (4 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-791-2
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L
- Cat. No.:78533-1L
- Miljøcertificering:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø