Orders LinkContactUsLinkComponent
2,4-Dihydroxy-5-fluorpyrimidin
2,4-Dihydroxy-5-fluorpyrimidin
Toxic
Health
Supplier:   Cayman Chemical
2,4-Dihydroxy-5-fluorpyrimidin
Supplier:  Cayman Chemical
Certificates
2,4-Dihydroxy-5-fluorpyrimidin
4 Options Available Below
Product Details & Documents
Synonyms:  Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU
  • CAS Number:51-21-8
  • Formula:C₄H₃FN₂O₂
  • MDL Number:MFCD00006018
  • Molecular Weight:130.08 g/mol
  • Merck Index:13,04208
  • Boiling Pt:347 °C (1013 hPa)
  • Melting Pt:282…283 °C
  • Storage Temperature:Stue temperatur
  • EINECS:200-085-6
  • UN:2811
  • ADR:6.1,III
Product Family Options
Product Information
  • Pakning
    Pakn
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 592,00 kr
    592,00 kr
    1 * 5 g
       592,00 kr
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 1.140,00 kr
    1.140,00 kr
    1 * 10 g
       1.140,00 kr
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 1.930,00 kr
    1.930,00 kr
    1 * 25 g
       1.930,00 kr
  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 3.560,00 kr
    3.560,00 kr
    1 * 50 g
       3.560,00 kr