Orders LinkContactUsLinkComponent
Spectrophotometer cuvettes
 :   EPPENDORF
Spectrophotometer cuvettes
 :  EPPENDORF
Spectrophotometer cuvettes
Spectrophotometer cuvettes
Spectrophotometer cuvettes suitable for colorimetric protein assays, kinetic and fluorescence measurements.
  • Catalog No:
  • 634-0833
  • 634-0832
  • Environmentally Preferable:
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Miljøcertificering:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Product Family Options
  • Model
    Område
    Højde
    Volumen
    Længde på lysvej
    Vinduesmateriale
  • Semi-micro
    300 − 900 nm
    45 mm
    3 ml
    10 mm
    PMMA
    1 * 1.000 Items
    1 * 1.000 Items - 1.110,00 kr
    1.110,00 kr
    1 * 1.000 Items
       1.110,00 kr
      Lav fremstillingspåvirkning
    Specifications
    Cat. No.
    634-0833
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • Macro
    300 − 900 nm
    45 mm
    4,5 ml
    10 mm
    PMMA
    1 * 1.000 Items
    1 * 1.000 Items - 1.050,00 kr
    1.050,00 kr
    1 * 1.000 Items
       1.050,00 kr
      Lav fremstillingspåvirkning
    Specifications
    Cat. No.
    634-0832
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001