Orders LinkContactUsLinkComponent
Repeating adapter for syringes
Repeating adapter for syringes
RAX repeating adapter, for syringes from 500 nl - 500 μl
Katalog # SGEA031930
Repeating adapter for syringes
Katalog # SGEA031930
Repeating adapter for syringes
RAX repeating adapter, for syringes from 500 nl - 500 μl
Price
614,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The RAX repeating adapter improves precision and reproducibility when repeatedly dispensing the same volume manually. It is suitable for syringes (except those with plunger stops, or diluter/dispenser syringes) and can be used on syringe barrels (Ext.Ø 6,5 and 8 mm).
  • Easy to use finger grips and can be set to any desired volume
  • Used as added protection against both plunger blow out at elevated pressures and plunger bending
The flat sides allow it to sit securely on the bench with the needle in a raised position helping to prevent damage from rolling. The double-ended tool facilitates assembly, it can easily and quickly be installed and if required it can readily be removed, replaced or swapped between syringes.
Leveringsinformation: Supplied with double-ended tool.
Specifikationer
  • Beskrivelse:
    RAX repeating adapter, for syringes from 500 nl - 500 μl