Orders LinkContactUsLinkComponent
Pipette controller, Pipet-Aid®
Pipette controller, Pipet-Aid®
Pipet-Aid, with dual pump filtration unit, 220 V, EU-plug
Katalog # DRUM4-000-221
Leverandør:   DRUMMOND
Pipette controller, Pipet-Aid®
Katalog # DRUM4-000-221
Leverandør:  DRUMMOND
Pipette controller, Pipet-Aid®
Pipette controller, Pipet-Aid®
Pipet-Aid, with dual pump filtration unit, 220 V, EU-plug
Price
8.060,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This controller with easy-to-use volume control mechanism permits precise fluid intake and delivery. Can be used with glass or plastic pipettes.
  • Check valve protects against accidental fluid intake into the handle and
  • Special filtration unit for use with biohazardous materials is available
  • Dual pump filtration unit has aretention efficiency of 99,9999+% for 0,1 µm particles and droplets
  • Optional Tissue Culture (TC) nosepiece will prevent cross-contamination due to over-pipetting
Specifikationer
  • Beskrivelse:
    Pipet-Aid, with dual pump filtration unit, 220 V, EU-plug
  • Seq:
    04