Reagent containers for Microlab® STAR line
Om denne vare
PP, white or black. These reagent reservoirs with 60, 120 or 200 ml capacity can be placed in a reagent carrier on the STAR line instruments. The reservoirs are available self-standing or non-self-standing.
- Special bottom geometry minimises residual volumes
- With volume markings, without or with lid
- With one (60 ml), three (120 ml) or four (200 ml) wave breakers to avoid splashes during movements of the filled containers
- IVD compliant
- Autoclavable
Ordering information: The carriers are not included and need to be ordered separately.
6 tilgængelige muligheder nedenfor
Product Details & Documents
PP, white or black. These reagent reservoirs with 60, 120 or 200 ml capacity can be placed in a reagent carrier on the STAR line instruments. The reservoirs are available self-standing or non-self-standing.
- Special bottom geometry minimises residual volumes
- With volume markings, without or with lid
- With one (60 ml), three (120 ml) or four (200 ml) wave breakers to avoid splashes during movements of the filled containers
- IVD compliant
- Autoclavable
Ordering information: The carriers are not included and need to be ordered separately.
Documents
Product Family Options
Produktinformation
- ModelKapacitetB×D×HFarvePakketTilgængelighedPrice
- 736-0830Reagent container without lid, self-standing, 1TReagent container without lid, self-standing, 1TSelf-standing, 1T60 ml89,7×20,0×64,8 mmtransparentIndividually wrappedLog ind for tilgængelighed1 * 28 Items1.051,06 krSpecifikationer:MarketsourceMarketsource
- 736-0831Reagent container with lid, self-standing, 1TReagent container with lid, self-standing, 1TSelf-standing, 1T, with lid60 ml89,7×20,0×64,8 mmgennemsigtigIndividually wrappedLog ind for tilgængelighed1 * 28 Items1.051,06 krSpecifikationer:MarketsourceMarketsource
- 736-0832Reagent container with lid, self-standing, 1T, black, for light sensitive samplesReagent container with lid, self-standing, 1T, black, for light sensitive samplesSelf-standing, black, 1T, with lid60 ml89,7×20,0×64,8 mmsortIndividually wrappedLog ind for tilgængelighed1 * 28 ItemsSpecifikationer:MarketsourceMarketsource
- HAMR187297Reagent container without lid, non self-standingReagent container without lid, non self-standingNon self-standing50 ml90×20×60 mmgennemsigtigIndividually wrappedLog ind for tilgængelighed1 * 12 ItemsSpecifikationer:MarketsourceMarketsource
- 736-0834Reagent container without lid, self-standing, 1TReagent container without lid, self-standing, 1TSelf-standing, 1T120 ml142,8×19,1×80,0 mmgennemsigtigIndividually wrappedLog ind for tilgængelighed1 * 12 Items684,03 krSpecifikationer:MarketsourceMarketsource
- 736-0833Reagent container without lid, non self-standing, 1TReagent container without lid, non self-standing, 1TNon self-standing, 1T120 ml155,9×21,1×91,5 mmgennemsigtigIndividually wrappedLog ind for tilgængelighed1 * 12 Items684,03 krSpecifikationer:MarketsourceMarketsource