5,8-Dibromisoquinolin
Glass bottle for solids, 1 g
Katalog # ACTIAS1278G1
Marketsource
Leverandør: ACTIVATE SCIENTIFIC
5,8-Dibromisoquinolin
Katalog # ACTIAS1278G1
Leverandør: ACTIVATE SCIENTIFIC
Marketsource
CAS nummer: 81045-39-8
Gå til:
Product Details & Documents
- CAS nummer:81045-39-8
- Formel:C₉H₅Br₂N
- MDL nummer:MFCD03109888
- Molecular Weight:286.95 g/mol
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 g



